missing translation for 'onlineSavingsMsg'
Learn More

UNC0638, MedChemExpress

Product Code. 16456706
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16456706 100 mg 100mg
16436418 10 mg 10mg
16419007 5 mg 5mg
16402527 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16456706

Brand: Medchem Express HY15273

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC0638 selectively inhibits G9a and GLP histone methyltransferase activity with IC50s of less than 15 nM and 19 nM, respectively. UNC0638 has anti-FMDV (foot-and-mouth disease virus) and anti-VSV (vesicular stomatitis virus) activities.

UNC0638 selectively inhibits G9a and GLP histone methyltransferase activity with IC50s of less than 15 nM and 19 nM, respectively. UNC0638 has anti-FMDV (foot-and-mouth disease virus) and anti-VSV (vesicular stomatitis virus) activities.

Specifications

Chemical Name or Material UNC0638
CAS 1255580-76-7
Color White
Purity Grade Notes Research
Molecular Formula C30H47N5O2
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (58.85 mM)
SMILES CC(N1CCC(NC2=C3C=C(OC)C(OCCCN4CCCC4)=CC3=NC(C5CCCCC5)=N2)CC1)C
Molecular Weight (g/mol) 509.73
Formula Weight 509.73
Percent Purity 98.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.