missing translation for 'onlineSavingsMsg'
Learn More

UNC0224, MedChemExpress

Product Code. 16497007
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497007 5 mg 5mg
16490617 50 mg 50mg
16484288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497007 Supplier Medchem Express Supplier No. HY10929

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC0224 is a potent and selective histone methyltransferase G9a inhibitor with a Ki of 2.6 nM, an IC50 of 15 nM and a Kd of 23 nM. UNC0224 also potently inhibits b>GLP with assay-dependent IC50 values of 20-58 nM. UNC0224 is inactive against SET7/9, SET8/PreSET7, PRMT3 and JMJD2E.

UNC0224 is a potent and selective histone methyltransferase G9a inhibitor with a Ki of 2.6 nM, an IC50 of 15 nM and a Kd of 23 nM. UNC0224 also potently inhibits b>GLP with assay-dependent IC50 values of 20-58 nM. UNC0224 is inactive against SET7/9, SET8/PreSET7, PRMT3 and JMJD2E.

Specifications

Chemical Name or Material UNC0224
CAS 1197196-48-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H43N7O2
Quantity 5 mg
Solubility Information DMSO : 16.67 mg/mL (34.32 mM; Need ultrasonic)
SMILES CN1CCC(NC2=C3C=C(OC)C(OCCCN(C)C)=CC3=NC(N4CCN(C)CCC4)=N2)CC1
Molecular Weight (g/mol) 485.67
Formula Weight 485.67
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.