missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UNC 0224
Description
Potent G9a and GLP inhibitor (IC50 values are 15 and 20 nM in a ThioGlo assay, respectively). Exhibits >1000-fold selectivity for G9a over SET7, SET8 and SET9.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 485.67 |
| Purity | >98% |
| Molecular Weight (g/mol) | 485.677 |
| Chemical Name or Material | UNC 0224 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?