missing translation for 'onlineSavingsMsg'
Learn More

Merck Ultrafiltration Discs, 300 kDa NMW, Biomax Polyethersulfone

Product Code. 10035371 Shop All Merck Products
Change view
Click to view available options
Diameter (Metric) Filter:
25 mm
44.5 mm
47 mm
63.5 mm
76 mm
90 mm
150 mm
Unit Size:
Pack of 10
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Filter unitSize
10035371 90 mm Pack of 5
10711991 25 mm Pack of 10
10025291 44.5 mm Pack of 10
10641412 47 mm Pack of 10
10719922 63.5 mm Pack of 10
10116761 76 mm Pack of 10
10515482 150 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10035371 Supplier Merck Supplier No. PBMK09005
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

300 kDa NMWL, Biomax polyethersulfone

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.

Features & Benefits:

  • Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
  • Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements
  • Compatible with Amicon™ Stirred Cells

Applications:
Concentration, Buffer Exchange, Desalting

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Material PES
MWCO 300 kDa
Filter Code PBMK
Filtration Area 63 cm2
For Use With (Application) Protein purification; protein concentration; protein desalting
Housing Material PES
Quantity 5Pack
Type Ultrafiltration Disc
Diameter (Metric) 90mm
Diameter (Metric) Filter 90 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.