missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UCK2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10028829
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10028829 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10028829 Supplier Invitrogen™ Supplier No. PA514010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene UCK2
Gene Accession No. Q9BZX2
Gene Alias AA407809; AI481316; AU018180; AU020720; cytidine monophosphokinase 2; RGD:620742}; Testis-specific protein TSA903; TSA903; UCK 2; Uck2; uck2 {ECO:0000312; UK; UMK; UMPK; uridine kinase; uridine monophosphate kinase; uridine monophosphate kinase 2; uridine monophosphokinase 2; Uridine-cytidine kinase 2
Gene Symbols UCK2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human UCK2.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7371
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.