missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ubiquilin 2 Monoclonal Antibody (3B8A10)
GREENER_CHOICE

Product Code. 10179582
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10179582 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10179582 Supplier Invitrogen™ Supplier No. 354400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

Ubiquilin 2 Monoclonal Antibody for Western Blot, ICC/IF, ELISA

This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases; and thus, are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to bind the ATPase domain of the Hsp70-like Stch protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquilin 2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3B8A10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene UBQLN2
Gene Accession No. Q9UHD9
Gene Alias ALS15; CHAP1; DSK2; DSK2 homolog; HRIHFB2157; N4BP4; Nedd4 binding protein 4; Plic2; Plic-2; protein linking IAP with cytoskeleton 2; RGD1563566; ubiquilin 2; ubiquilin-2; Ubiquitin-like product Chap1/Dsk2; Ubiquitin-related protein; Ubqln2
Gene Symbols UBQLN2
Host Species Mouse
Immunogen Recombinant protein corresponding to the C-terminal region of the human ubiquilin protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29978
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.