Learn More
Macherey-Nagel™ Turmeric Qualitative Test Paper
Brand: Macherey-Nagel™ 90747
This item is not returnable.
View return policy
Specifications
| Yellow to Red | |
| Determination of Boric Acid and Borates | |
| Qualitative Test Paper |
| Strip | |
| 200 Pack |