Learn More
BOLA Tube Fittings, Elbow
Description
- Tube fitting elbow-shaped made of PTFE, two connections with nuts made of glass-fibre reinforced PTFE.
- Universal chemical resistance, the product is only exposed to PTFE.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Diameter (Metric) | 12 mm |
| Autoclavable | Autoclavable |
| Color | White |
| Flexibility | Rigid |