missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA Tube End Fittings
Description
- Tube end fittings.
- With male thread UNF ¼ in. 28 G washers made of PA are included in delivery.
- Packing unit: 10 pieces, differing ordering quantities are rounded up to factor 10.
Specifications
Specifications
| Color | Red |
| Diameter (Metric) Outer | 3.2 mm |
| Diameter (Metric) Inner | 1.6 mm |
| Autoclavable | Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?