Learn More
Description
The Thermo Scientific™ TSG Series general purpose laboratory refrigerators are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
TSG Series General Purpose Refrigerators are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
- Wide range of sizes from 400L to 1500L double door
- Glass door (triple pane) and solid door configurations
- Whisper quite operation < 50 dBA
- White painted interior and exterior
- Adaptive Defrost
- Temperature, door ajar, and power failure (with battery backup) alarms.
- Low heat output
- Rear access port
- Keyed Lock
- LED interior lighting
- Green hydrocarbon refrigerants
- Remote alarm contacts
Specifications
Specifications
| Description | TSG Series General Purpose Laboratory Refrigerator |
| Capacity (Metric) | 1500 L |
| Temperature Range | 2°C to 10°C |
| Temperature (Setpoint) | +5°C |
| Refrigerant | R290 |
| Door Style | Glass |
| No. of Doors | 2 |
| Shelves | 8 Adjustable |
| Capacity (Metric) Shelf | 30 kg |
| Cabinet Material | Steel |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

