missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1,10-phenanthroline)iron(II) sulfate, 0.025M aqueous solution
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Color | Red |
| Density | 1.0060g/mL |
| Packaging | Glass bottle |
| Quantity | 500 mL |
| Specific Gravity | 1.006 |
| Formula Weight | 692.53 |
| Physical Form | Liquid |
| Chemical Name or Material | Tris(1, 10-phenanthroline)iron(II) sulfate, 0.025M Aqueous Solution |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction