missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethyl mercaptan, 97%

Product Code. 10127603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127603 500 g Glass bottle 500g
10051620 25 g Glass bottle 25g
10235000 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127603 Supplier Thermo Scientific Acros Supplier No. 140415000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3695-77-0 | C19H16S | 276.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3695-77-0
Molecular Formula C19H16S
Molecular Weight (g/mol) 276.39
MDL Number MFCD00004854
InChI Key JQZIKLPHXXBMCA-UHFFFAOYSA-N
Synonym triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
PubChem CID 77281
IUPAC Name triphenylmethanethiol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S

Specifications

Melting Point 103.0°C to 107.0°C
Color Cream to Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3CSH
Quantity 500 g
Beilstein 06, I, 352
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 276.39
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Triphenylmethyl mercaptan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.