missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium hydroxide, 0.25M solution in methanol

Product Code. 10036222 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10036222 10 mL 10mL
10516353 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10036222 Supplier Thermo Scientific Acros Supplier No. 381360100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17287-03-5 | C3H9IS | 204.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trimethylsulfonium hydroxide
Color Colorless
Name Note 0.25M solution in methanol
CAS 17287-03-5, 67-56-1
Molecular Formula C3H9IS
MDL Number MFCD00011632
Concentration 0.2 to 0.3M
Density 0.81g/mL
Synonym trimethylsulfonium hydroxide, trimethylsulfonium hydroxide solution, trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent, sulfonium, trimethyl-, hydroxide, acmc-209sky, trimethylsulfoniumhydroxyd, ksc498e2f, trimethylsulfoniumhydroxidesolution, trimethylsulfonium hydroxide, 0.2mol/l methanol solution, trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent
InChI Key VFJYIHQDILEQNR-UHFFFAOYSA-M
SMILES [I-].C[S+](C)C
IUPAC Name trimethylsulfanium;hydroxide
Molecular Weight (g/mol) 204.07
PubChem CID 11105313
Formula Weight 94.18
Packaging Glass bottle
Quantity 10 mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylsulfonium hydroxide, 0.25M solution in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.