missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™

Product Code. 10183064 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10183064 800 mL 800mL
10366654 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10183064 Supplier Thermo Scientific Acros Supplier No. 377458000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1822-00-0 | C4H11LiSi | 94.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Trimethylsilyl)methyllithium
Name Note 0.8M (12 wt%) solution in hexanes
CAS 1822-00-0, 92112-69-1, 92112-69-1
CAS Min % 7.0, 86.0
CAS Max % 14.0, 93.0
Color Brown-Yellow to Colorless
Density 0.6500g/mL
Boiling Point 65.0°C to 70.0°C
Flash Point −40°C
Packaging AcroSeal glass Bottle packed in tin can
Molecular Formula C4H11LiSi
Linear Formula LiCH2Si(CH3)3
MDL Number MFCD00010747
Quantity 800 mL
Specific Gravity 0.65
Synonym trimethylsilyl methyllithium, trimethylsilyl methyl lithium, lich2sime3, pubchem18144, tms-ch2li, lithiomethyltrimethylsilane, acmc-1bvf0, trimethylsilylmethyl lithium
InChI Key KVWLUDFGXDFFON-UHFFFAOYSA-N
SMILES [Li+].C[Si](C)(C)[CH2-]
IUPAC Name lithium;methanidyl(trimethyl)silane
Molecular Weight (g/mol) 94.16
PubChem CID 3482579
Formula Weight 94.16
Percent Purity 7 to 14 wt%
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.