missing translation for 'onlineSavingsMsg'
Learn More

Trifludimoxazin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30314186
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30314186 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30314186

Brand: Dr. Ehrenstorfer DREA17843300AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifludimoxazin
CAS 1258836-72-4
Analyte or Component Names Trifludimoxazin
Shipping Condition +20°C
Molecular Formula C16 H11 F3 N4 O4 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Dihydro-1,5-dimethyl-6-thioxo-3-[2,2,7-trifluoro-3,4-dihydro-3-oxo-4-(2-propyn-1-yl)-2H-1,4-benzoxazin-6-yl]-1,3,5-triazine-2,4(1H,3H)-dione, BAS 850H, Trifludimoxazin
InChI Formula InChI=1S/C16H11F3N4O4S/c1-4-5-22-10-7-9(8(17)6-11(10)27-16(18,19)12(22)24)23-13(25)20(2)15(28)21(3)14(23)26/h1,6-7H,5H2,2-3H3
SMILES CN1C(=O)N(C(=O)N(C)C1=S)c2cc3N(CC#C)C(=O)C(F)(F)Oc3cc2F
IUPAC Name 1,5-dimethyl-6-sulfanylidene-3-(2,2,7-trifluoro-3-oxo-4-prop-2-ynyl-1,4-benzoxazin-6-yl)-1,3,5-triazinane-2,4-dione
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 412.34
Quantity 1 mL
Formula Weight 412.0453 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.