missing translation for 'onlineSavingsMsg'
Learn More

α-tri-Calcium Phosphate, puriss. p.a., ≥75% α to Phase Basis, Honeywell Fluka™

Product Code. p-7120002 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Poly Bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15625770 10 g Poly Bottle 10g
15615770 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15625770 Supplier Honeywell-Fluka Supplier No. 5373710G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥75% α-phase basis (sinterted powder)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-87-4
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.174
MDL Number MFCD00015984
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
PubChem CID 24456
ChEBI CHEBI:9679
IUPAC Name tricalcium;diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]

Specifications

Melting Point 1670°C
Quantity 10 g
UN Number NONH for all modes of transport
Formula Weight 310.18g/mol
Percent Purity ≥75% α to phase basis (sintered powder)
Packaging Poly Bottle
Appearance Sintered powder
Density 3.27g/cm3
Chemical Name or Material α-tri-Calcium phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.