missing translation for 'onlineSavingsMsg'
Learn More

trans-Loperamide N-Oxide, TRC

Product Code. 30136571
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136571 100 mg 100mg
30134983 5 mg 5mg
18062820 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136571 Supplier Toronto Research Chemicals Supplier No. TRCL469460100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106900-12-3
Molecular Formula C29 H33 Cl N2 O3
Molecular Weight (g/mol) 493.04
Synonym Loperamide Oxide, Loperamide Hydrochloride Imp. F (EP), trans-4-(4-chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutanamide 1-oxide, Arestal, Loperamide oxide, R 58425
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxy-1-oxidopiperidin-1-ium-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide
SMILES CN(C)C(=O)C(CC[N@+]1([O-])CC[C@](O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C29H33ClN2O3/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32(35)20-17-28(34,18-21-32)23-13-15-26(30)16-14-23/h3-16,34H,17-22H2,1-2H3/t28-,32-
Formula Weight 492.218
Percent Purity >95%
Chemical Name or Material Loperamide Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.