missing translation for 'onlineSavingsMsg'
Learn More

trans-Benzyl(chloro)bis(triphenylphosphine)palladium(II)

Product Code. 10150812 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10150812 500 mg Glass bottle 500mg
10552242 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10150812 Supplier Thermo Scientific Acros Supplier No. 369715000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22784-59-4 | C43H37ClP2Pd | 757.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22784-59-4
Molecular Formula C43H37ClP2Pd
Molecular Weight (g/mol) 757.59
MDL Number MFCD00010395
InChI Key GYAXNTWMUGUEJY-UHFFFAOYSA-M
IUPAC Name benzyl(chloro)palladiumbis(ylium); bis(triphenylphosphane)
SMILES Cl[Pd++]CC1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 166°C to 170°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 mg
Fieser 08,35; 09,41; 10,26; 12,44; 13,30
Formula Weight 757.59
Physical Form Crystalline Powder
Chemical Name or Material trans-Benzyl(chloro)bis(triphenylphosphine)palladium(II)
compliance-icons
Product Identifier
  • trans-Benzyl(chloro)bis(triphenylphosphine)palladium(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.