missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentene, 99%

Product Code. 11367416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367416 1 g 1g
11377416 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367416 Supplier Thermo Scientific Alfa Aesar Supplier No. L09421.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 646-04-8 | C5H10 | 70.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-04-8
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00009384
InChI Key QMMOXUPEWRXHJS-HWKANZROSA-N
Synonym trans-2-pentene, e-2-pentene, 2-pentene, 3-pentene, 2-pentene, e, sym-methylethylethylene, e-pent-2-ene, beta-n-amylene, trans-beta-amylene, 2-pentene, 2e
PubChem CID 5326161
IUPAC Name (E)-pent-2-ene
SMILES CCC=CC

Specifications

Melting Point -136°C
Boiling Point 36°C to 37°C
Flash Point −45°C (−49°F)
Odor Petroleum-like
Quantity 1 g
UN Number UN3295
Beilstein 1718795
Merck Index 14,7123
Formula Weight 70.14
Percent Purity 99%
Chemical Name or Material trans-2-Pentene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • trans-2-Pentene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: .beta.-Amylene-trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.