Learn More
Invitrogen™ TRA-1-81 Monoclonal Antibody (cl.26)
Description
TRA-1-81 recognizes an antigen of similar size detected as by TRA-1-60, GCTM2, K4, and K21, and all these antibodies detect similar but distinct patterns of expression on panels of tumor-derived cell lines. TRA-1-81 and TRA-1-60 epitopes are associated with distinct molecular species. TRA-1-81, TRA-1-60, GCTM2, and K21 antibodies all recognize distinct epitopes formed by modifications of a common keratan sulfate core molecule that is characteristically expressed by human EC cells. TRA-1-81, clone cl.26, likely recognizes the carbohydrate part of the molecule as TRA-1-60 activity is enhanced by neuraminidase. This antibody is specific for the TRA-1-81 antigen. On western blots, it identifies the target band at ~200-400 kDa. Reactivity has been confirmed with human EC1003.7 and NTERA-2 cell lysates by western blotting, with smooth muscle cells in frozen normal human arterioles and arteries, intestine, lung, skin, and uterus, as well as epithelial cells of the mammary ducts, stomach, small and large intestine tissues by indirect immunofluorescence, and with lysates from 125I surface-labeled 2102Ep cl.2A6 human EC cells by immunoprecipitation.
Specifications
Specifications
| Antigen | TRA-1-81 |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | cl.26 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene Alias | GCTM-2 antigen; Gp200; PC; PCLP; PCLP1; PCLP-1; Podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; PODXL; TRA180; tra1-81 |
| Host Species | Mouse |
| Immunogen | 2102Ep cl.2A6 human embryonal carcinoma cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.