missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOR4A Recombinant Rabbit Monoclonal Antibody (K01_1N76)
Description
TOR4A Recombinant Monoclonal Antibody for Western Blot
TOR4A gene ontology annotations related to this gene include platelet degranulation.
Specifications
Specifications
| Antigen | TOR4A |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1N76 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | TOR4A |
| Gene Accession No. | Q9NXH8 |
| Gene Alias | C9H9orf167; C9orf167; RGD1308019; TOR4A; Torsin family 4 member A; torsin family 4, member A; torsin family protein C9orf167; torsin-4A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?