missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOR4A Recombinant Rabbit Monoclonal Antibody (K01_1N76)
GREENER_CHOICE

Product Code. 30824621
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30824621 100 μL
30829655 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30824621 Supplier Invitrogen™ Supplier No. MA563204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TOR4A Recombinant Monoclonal Antibody for Western Blot

TOR4A gene ontology annotations related to this gene include platelet degranulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOR4A
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1N76
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene TOR4A
Gene Accession No. Q9NXH8
Gene Alias C9H9orf167; C9orf167; RGD1308019; TOR4A; Torsin family 4 member A; torsin family 4, member A; torsin family protein C9orf167; torsin-4A
Gene Symbols TOR4A
Host Species Rabbit
Immunogen Recombinant protein of human C9orf167.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54863
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.