missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.
- Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
- Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
- Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
- Performance is guaranteed through the expiration period stated on the label
- Suitable for exacting EPA sample cleanup protocols
Specifications
Specifications
| Melting Point | -95°C |
| Color | Colorless |
| Density | 0.87g/cm3 at 20°C |
| Boiling Point | 110°C to 111°C |
| Quantity | 1 L |
| Explosion Limit | 1.2 - 7 Vol % |
| Flash Point | 4°C |
| Ignition Point | 535°C |
| Odor | Aromatic |
| Packaging | Glass |
| Show More |
Safety and Handling
Signal Word
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?