missing translation for 'onlineSavingsMsg'
Learn More

Tirbanibulin Mesylate, MedChemExpress

Product Code. 16494876
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494876 100 mg 100mg
16456567 5 mg 5mg
16430217 50 mg 50mg
16403848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494876 Supplier Medchem Express Supplier No. HY10340B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.

Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.

Specifications

Chemical Name or Material Tirbanibulin Mesylate
CAS 1080645-95-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H33N3O6S
Quantity 100 mg
Synonym KX2-391 MesylateKX01 Mesylate
Solubility Information DMSO : 100 mg/mL (189.53 mM; Need ultrasonic)
SMILES O=C(NCC1=CC=CC=C1)CC2=NC=C(C3=CC=C(OCCN4CCOCC4)C=C3)C=C2.CS(=O)(O)=O
Molecular Weight (g/mol) 527.63
Formula Weight 527.63
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.