missing translation for 'onlineSavingsMsg'
Learn More

Tirbanibulin dihydrochloride, MedChemExpress

Product Code. p-200097038
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493838 10 mg 10mg
16484876 100 mg 100mg
16446567 5 mg 5mg
16420217 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493838 Supplier Medchem Express Supplier No. HY10340A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tirbanibulin (dihydrochloride) (KX2-391 (dihydrochloride)) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.

Tirbanibulin (dihydrochloride) (KX2-391 (dihydrochloride)) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.

Specifications

Chemical Name or Material Tirbanibulin dihydrochloride
CAS 1038395-65-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H31Cl2N3O3
Quantity 10 mg
Synonym KX2-391 dihydrochlorideKX-01 dihydrochloride
Solubility Information H2O : 50 mg/mL (99.12 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (66.07 mM; Need ultrasonic)
SMILES O=C(CC1=NC=C(C2=CC=C(OCCN3CCOCC3)C=C2)C=C1)NCC4=CC=CC=C4.Cl.Cl
Molecular Weight (g/mol) 504.45
Formula Weight 504.45
Percent Purity 96.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.