missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tirbanibulin dihydrochloride, MedChemExpress
Description
Tirbanibulin (dihydrochloride) (KX2-391 (dihydrochloride)) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.
Specifications
Specifications
| Chemical Name or Material | Tirbanibulin dihydrochloride |
| CAS | 1038395-65-1 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H31Cl2N3O3 |
| Quantity | 10 mg |
| Synonym | KX2-391 dihydrochlorideKX-01 dihydrochloride |
| Solubility Information | H2O : 50 mg/mL (99.12 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (66.07 mM; Need ultrasonic) |
| SMILES | O=C(CC1=NC=C(C2=CC=C(OCCN3CCOCC3)C=C2)C=C1)NCC4=CC=CC=C4.Cl.Cl |
| Molecular Weight (g/mol) | 504.45 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?