missing translation for 'onlineSavingsMsg'
Learn More

Tipepidine Hibenzate, TRC

Product Code. 30452549
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452549 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452549 Supplier Toronto Research Chemicals Supplier No. TRCT31139050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tipepidine Hibenzate
CAS 31139-87-4
Molecular Formula C15 H17 N S2 . C14 H10 O4
Quantity 50 mg
Synonym Benzoic acid','2-(4-hydroxybenzoyl)-','compd. with 3-(di-2-thienylmethylene)-1-methylpiperidine (1:1), Benzoic acid','o-(p-hydroxybenzoyl)-','compd. with 3-(di-2-thienylmethylene)-1-methylpiperidine (1:1) (8CI), Benzoic acid','o-(p-hydroxybenzoyl)-','compd. wit
InChI Formula InChI=1S/C15H17NS2.C14H10O4/c1-16-8-2-5-12(11-16)15(13-6-3-9-17-13)14-7-4-10-18-14;15-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)14(17)18/h3-4,6-7,9-10H,2,5,8,11H2,1H3;1-8,15H,(H,17,18)
SMILES CN1CCCC(=C(c2cccs2)c3cccs3)C1.OC(=O)c4ccccc4C(=O)c5ccc(O)cc5
IUPAC Name 3-(dithiophen-2-ylmethylidene)-1-methylpiperidine;2-(4-hydroxybenzoyl)benzoic acid
Molecular Weight (g/mol) 517.659
Formula Weight 517.138
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.