Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator with Standard Optics
Product Code. 10161083
Change view
Click to view available options
:
Comprator with Standard Optics
Product Code.
10161083 Comprator with Standard Optics
1 options
Product Code. 10161083 Supplier Lovibond™ Supplier No. 145000
Only null left
Add to basket
Add to basket
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
  • The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
  • Suitable for both mobile and static water analysis
  • Available with a number of different color scales
  • Provides the basis for a comprehensive easy to to to use analysis system
  • Each Checkit™ Comparator Disc contains a continuous colour scale for exact color matching between standard and sample
  • Each Checkit™ Comparator Kit is supplied in a carry case with comparator, vials, discs, reagents and method instructions for all tests
Specifications
Description Dia.: 13.5mm
For Use With (Application) Water Quality Testing
For Use With (Equipment) Checkit Comprator Tests
Includes Carry case with comparator, vials, discs, reagents and method instructions for all tests
Type Checkit Comprator with Standard Optics