missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Molybdate LR Test, 0 to 10mg/L
Description
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous color scale for exact color matching between standard and sample
- The D55 Checkit Comparator contains mirror optics which extend the cell path length to 55mm, allowing greater accuracy at lower concentrations
- Checkit™ Comparator Copper LR Kit is supplied in a carry case with D55 Comparator, Vials, Discs, reagents and method instructions for all tests
- The Molybdate disc supplied measures from 0 to 10mg/L with the following steps: 0 / 1 / 2 / 3 / 4 / 5 / 6 / 7 / 8 / 9 / 10
Specifications
Specifications
| For Use With (Application) | Water Quality Testing |
| For Use With (Equipment) | Checkit™ Comparator D55 |
| Includes | Carry case with D55 Comparator, Vials, Discs, reagents and method instructions |
| Range | 0 to 10mg/L |
| Type | Molybdate LR Test Disc for Checkit™ Comprator D55 |
Safety and Handling
Product Identifier
- Comparator Checkit, Molybdat LR, 0-10,0mg/l MoO4 .
- Danger
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Adipic acid, Sodiumbisulphate, Potassium persulphate, Calcium thioglycolate trihydrate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?