missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride, 1M solution in dichloromethane, AcroSeal™

Product Code. 10055992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055992 800 mL AcroSeal™ Glass Bottle 800mL
10629784 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055992 Supplier Thermo Scientific Acros Supplier No. undefined
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7646-78-8 | Cl4Sn | 260.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tin(IV) chloride
Name Note 1M Solution in Dichloromethane
CAS 7646-78-8, 75-09-2
Quantity 800 mL
Molecular Formula Cl4Sn
MDL Number MFCD00011242
Fieser 09,436; 17,335; 14,304; 13,300; 12,486; 10,370; 07,342; 06,553; 05,627; 03,269; 01,1111; 11,552
Merck Index 15, 8901
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
Solubility Information Solubility in water: reacts
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
SMILES Cl[Sn](Cl)(Cl)Cl
IUPAC Name tetrachlorostannane
Molecular Weight (g/mol) 260.52
PubChem CID 24287
Formula Weight 260.52
Concentration or Composition (by Analyte or Components) 1M, exact strength on the certificate of analysis
Packaging AcroSeal™ Glass Bottle
Appearance Clear colorless solution, fuming in air
Specific Gravity 1.419
Density 1.4190g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tin(IV) chloride, 1M solution in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.