Learn More
Invitrogen™ Thrombospondin 1 Recombinant Rabbit Monoclonal Antibody (K01_2P75)

Description
Immunogen sequence: PIDGCLSNPC FAGVKCTSYP DGSWKCGACP PGYSGNGIQC TDVDECKEVP DACFNHNGEH RCENTDPGYN CLPCPPRFTG SQPFGQGVEH ATANKQVCKP RNPCTDGTHD CNKNAKCNYL GHYSDPMYRC ECKPGYAGNG IICGEDTDLD GWPNENLVCV ANATYHCKKD NCPNLPNSGQ EDYDKDGIGD ACDDDDDNDK IPDDRDNCPF HYNPAQYDYD RDDVGDRCDN CPYNHNPDQA DTDNNGEGDA CAADIDGDGI LNERDNCQYV YNVDQRDTDM DGVGDQCDNC PLEHNPDQLD SDSDRIGDTC DNNQDIDEDG HQNNLDNCPY VPNANQADHD KDGKGDACDH DDDNDGIPDD KDNCRLVPNP DQKDSDGDGR GDACKDDFDH DSVPDIDDIC PENVDISETD FRRFQMIPLD PKGTSQNDPN WVVRHQGKEL VQTVNCDPGL AVGYDEFNAV DFSGTFFINT ERDDDYAGFV FGYQSSSRFY VVMWKQVTQS YWDTNPTRAQ GYSGLSVKVV NSTTGPGEHL RNALWHTGNT PGQVRTLWHD PRHIGWKDFT AYRWRLSHRP KTGFIRVVMY EGKKIMADSG PIYDKTYAGG RLGLFVFSQE MVFFSDLKYE CRDP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 54%, Pig - 99%, Cynomolgus monkey - 99%.
Specifications
Specifications
| Antigen | Thrombospondin 1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2P75 |
| Concentration | 0.60 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | THBS1 |
| Gene Accession No. | P07996, P35441 |
| Gene Alias | glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.