missing translation for 'onlineSavingsMsg'
Learn More

Thioflavin T

Product Code. p-200161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10589063 5 g Glass bottle 5g
10792631 25 g Glass bottle 25g
10005823 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10589063 Supplier Thermo Scientific Acros Supplier No. 211760050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2390-54-7 | C17H19ClN2S | 318.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-54-7
Molecular Formula C17H19ClN2S
Molecular Weight (g/mol) 318.86
MDL Number MFCD00011944
InChI Key JADVWWSKYZXRGX-UHFFFAOYSA-M
Synonym Basic Yellow 1, C.I. 49005
PubChem CID 16953
ChEBI CHEBI:76023
IUPAC Name 2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
SMILES [Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1

Specifications

Beilstein 27, 377
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most organic solvent
Infrared Spectrum Authentic
Formula Weight 318.85
Packaging Glass bottle
UV Spectrometry Complies
Color Ochre/Yellow to Yellow
Quantity 5 g
Physical Form Powder
Chemical Name or Material Thioflavin T

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.