missing translation for 'onlineSavingsMsg'
Learn More

Thiazol-5-ylmethyl N-((2R,5R)-5-Amino-1,6-diphenylhexan-2-yl)carbamate Hydrochloride, Mikromol™

Product Code. 30051102
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051102 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051102 Supplier Mikromol Supplier No. MM3623.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Thiazol-5-ylmethyl N-((2R,5R)-5-Amino-1,6-diphenylhexan-2-yl)carbamate Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiazol-5-ylmethyl N-((2R,5R)-5-Amino-1,6-diphenylhexan-2-yl)carbamate Hydrochloride
CAS 1247119-33-0
Analyte or Component Names Thiazol-5-ylmethyl N-((2R,5R)-5-Amino-1,6-diphenylhexan-2-yl)carbamate Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C23 H27 N3 O2 S . H Cl
Certifications/Compliance ISO 9001
Synonym Carbamic acid, N-[(1R,4R)-4-amino-5-phenyl-1-(phenylmethyl)pentyl]-, 5-thiazolylmethyl ester, hydrochloride (1:1), Thiazol-5-ylmethyl N-((1R,4R)-4-Amino-1-benzyl-5-phenylpentyl)carbamate Hydrochloride
InChI Formula InChI=1S/C23H27N3O2S.ClH/c24-20(13-18-7-3-1-4-8-18)11-12-21(14-19-9-5-2-6-10-19)26-23(27)28-16-22-15-25-17-29-22;/h1-10,15,17,20-21H,11-14,16,24H2,(H,26,27);1H/t20-,21-;/m1./s1
SMILES Cl.N[C@H](CC[C@H](Cc1ccccc1)NC(=O)OCc2cncs2)Cc3ccccc3
IUPAC Name thiazol-5-ylmethyl N-[(1R,4R)-4-amino-1-benzyl-5-phenyl-pentyl]carbamate;hydrochloride
Molecular Weight (g/mol) 446
Quantity 25 mg
Formula Weight 445.1591 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.