missing translation for 'onlineSavingsMsg'
Learn More

Theophylline-d6, TRC

Product Code. 30451769
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451769 2.5 mg 2.5mg
30450987 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30451769 Supplier Toronto Research Chemicals Supplier No. TRCT3438532.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Theophilline-D6
CAS 117490-39-8
Purity Grade Notes HPLC
Molecular Formula C7 2H6 H2 N4 O2
Quantity 2.5 mg
Synonym Theophylline-d6
InChI Formula InChI=1S/C7H8N4O2/c1-10-5-4(8-3-9-5)6(12)11(2)7(10)13/h3H,1-2H3,(H,8,9)/i1D3,2D3
SMILES [2H]C([2H])([2H])N1C(=O)N(c2nc[nH]c2C1=O)C([2H])([2H])[2H]
IUPAC Name 1,3-bis(trideuteriomethyl)-7H-purine-2,6-dione
Molecular Weight (g/mol) 186.2
Formula Weight 186.1024
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.