missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ebro Electronic™ TFX 430 Precision Thermometer

Description
TFX 430 Model Precision Thermometer. Powered by a replaceable lithium battery, these precision thermometers are handy, watertight temperature measurement devices for a whole range of measurement and control applications in the laboratory and in industry.
Specifications
Specifications
| Certifications/Compliance | EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485 |
| Diameter (Metric) Probe | 8 mm |
| Disposable | Single-use |
| For Use With (Application) | Applications: Laboratory, temperature monitoring, measure air, fluids, ductile substances, aggressive media and surfaces, Process monitoring |
| Height (Metric) | 54 mm |
| Includes | Thermometer, sensor, calibration certificate, instruction manual |
| Length (Metric) | 355 mm |
| Length (Metric) Probe | 190 mm |
| Material | Stainless Steel, Plastic |
| Model | TFX 430 and TPX 330 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?