missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TFN 520 / 530 Robust Thermocouple Thermometer Series
GREENER_CHOICE

Product Code. 10094100
Change view
Click to view available options
Product Type:
1-channel Thermometer
2-channel Thermometer
Probe Type:
Lemosa Connector
SMP Connector
IP Rating:
IP40
IP52
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Probe Type IP Rating unitSize
10094100 1-channel Thermometer Lemosa Connector IP52 Each
10155552 1-channel Thermometer SMP Connector IP40 Each
10389282 2-channel Thermometer SMP Connector IP40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10094100 Supplier Ebro Electronic™ Supplier No. 13405520A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High accurate thermometers for a whole range of measurement and control applications in the laboratory and in industry.

Temp. Range: -200 to 1200°C (K), -100 to 800°C (J), -50 to 300°C (T), -100 to 600°C (E)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012
Disposable Single-use
For Use With (Application) Applications: Heating technology, Air-conditioning, ventilation, Air, gases, Fluids, Motors, engines, Plastic materials, Laboratory
Height (Metric) 38 mm
Includes Thermometer, calibration certificate, instruction manual
Length (Metric) 190 mm
Material Plastic
Model TFN 520
Probe Type Lemosa Connector
Resolution 0.1°C at −200° to +250°C (K); 0.1°C at −100° to +190°C (J); 0.1°C at −50° to +220°C (T); 0.1°C at −100° to +150°C (E); 1°C for the rest
Weight (Metric) 0.3 kg
Product Type 1-channel Thermometer
Data Storage None
Max. Temperature (English) 2192°F
Max. Temperature (Metric) 1200°C
Min. Temperature (English) -328°F
Min. Temperature (Metric) -200°C
Temperature Range (Metric) -200°C to +1200°C (K), -100°C to +800°C (J), -50°C to +300°C (T), -100°C to +600°C (E)
Calibrated Yes
For Use With (Equipment) TPN probe series with lemosa connector, AG 130, AG 140, AN 140, AN 150
Battery Life Up to 5 years
Battery Type Lithium 3 V/Typ CR 2477
Accuracy ±0.3°C
Display LCD
Width (Metric) 120 mm
IP Rating IP52
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.