missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Texas Red Polyclonal Antibody
GREENER_CHOICE

Product Code. 10194282
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10194282 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10194282 Supplier Invitrogen™ Supplier No. A6399

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

We offer a variety of anti-fluorescent dye antibodies that recognize specific fluorophores and, in most cases, quench their fluorescence. These anti-dye antibodies, including those that recognize the tetramethylrhodamine (TRITC) and Texas Red™ fluorophores, can also serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria, or other biomolecules have been internalized by endocytic or pinocytic processes. Our anti-tetramethylrhodamine (A6397) and anti-Texas Red (A6399) antibodies have been raised against the tetramethylrhodamine and Texas Red fluorophores, respectively. These antibodies quench much of the fluorescence of the complementary dyes. However, due to the related chemical structures, the anti-tetramethylrhodamine antibody crossreacts with the Texas Red fluorophore and the anti-Texas Red antibody cross-reacts with tetramethylrhodamine. Also, both antibodies tightly bind the Rhodamine Red™ fluorophore. Thus, either antibody can serve as an effective probe for tetramethylrhodamine, Texas Red, or Rhodamine Red dyes. We use a sensitive quenching assay to ensure that this antibody is provided at a consistently high titer value. As supplied, 20 μL of the antibody solution is certified to produce ≥50% of the maximal fluorescence quenching of 1 mL of a 50 nM solution of Texas Red dye, assayed in 100 mM sodium phosphate, pH 8.0. Maximal quenching for Texas Red is ~60% of the fluorescence of the free dy...

Unlike biotin, which is an endogenous ligand in mitochondria, dye-based haptens permit background-free staining of cells and tissues and a sensitive alternative to biotin-streptavidin methods.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Texas Red
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias C31H29ClN2O6S2; Sulforhodamine
Host Species Rabbit
Immunogen Texas Red.
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.