missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water

Product Code. 10045553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045553 800 mL Glass bottle 800mL
10350444 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045553 Supplier Thermo Scientific Acros Supplier No. 389448000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-64-1 | C4H12ClO4P | 190.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrakis(hydroxymethyl)phosphonium chloride
CAS 124-64-1, 7732-18-5
Color Green-Yellow or Pink
Density 1.3400g/mL
Boiling Point 115.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H12ClO4P
MDL Number MFCD00031687
Quantity 800 mL
Specific Gravity 1.34
Synonym tetrakis hydroxymethyl phosphonium chloride, thpc, pyroset tkc, retardol c, tetramethylolphosphonium chloride, tetrakis hydroxymethyl phosphanium chloride, unii-58wb2xcf8i, proban cc, tetrakis hydroxymethyl phosphochloride, ccris 317
Solubility Information Solubility in water: soluble.
InChI Key AKXUUJCMWZFYMV-UHFFFAOYSA-M
SMILES [Cl-].OC[P+](CO)(CO)CO
IUPAC Name tetrakis(hydroxymethyl)phosphanium;chloride
Molecular Weight (g/mol) 190.56
PubChem CID 31298
Formula Weight 190.56
Percent Purity 80.0 to 85.0%
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrakis(hydroxymethyl)phosphonium chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H301-Toxic if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.