missing translation for 'onlineSavingsMsg'
Learn More

Tetracarbonyldi-μ-chlorodirhodium(I), 97%

Product Code. 10042580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042580 1 g Glass bottle 1g
10563561 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10042580 Supplier Thermo Scientific Acros Supplier No. 197600010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14523-22-9 | C4Cl2O4Rh2 | 388.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14523-22-9
Molecular Formula C4Cl2O4Rh2
Molecular Weight (g/mol) 388.75
MDL Number MFCD00135610
InChI Key FGKDXBICTVUSPK-UHFFFAOYSA-L
Synonym tetracarbonyldi-mu-chlorodirhodium i, rhodium carbonyl chloride i, rhodium i carbonyl chloride, rhodium i dicarbonyl chloride dimer, di-mu-chloro-tetracarbonyldirhodium i, chlorodicarbonylrhodium i dimer, 4co.cl2rh2, bis chlorodicarbonyl rhodium, tetracarbonyldi-, i-chlorodirhodium i, tetracarbonyldi-micron-chlorodirhodium i
IUPAC Name bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride
SMILES [Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Specifications

Melting Point 121.0°C
Color Brown to Red
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Rh)
Packaging Glass bottle
Linear Formula [Rh(CO)2Cl]2
Quantity 1 g
UN Number 2811
Merck Index 15, 8312
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents (except, aliphatic hydrocarbons)
Formula Weight 388.76
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Tetracarbonyldi-µ-chlorodirhodium(I)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.