missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)

Product Code. 10189919 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10189919 5 g Glass bottle 5g
10506250 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10189919 Supplier Thermo Scientific Acros Supplier No. 437000050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108374-32-9 | Cl2H12N4Pt | 334.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108374-32-9
Molecular Formula Cl2H12N4Pt
Molecular Weight (g/mol) 334.11
MDL Number MFCD00149947
InChI Key KHCPSOMSJYAQSY-UHFFFAOYSA-L
Synonym platinum 4+ ion hydrate hydrochloride tetraazanide
IUPAC Name platinum(2+) tetraamine dichloride
SMILES N.N.N.N.[Cl-].[Cl-].[Pt++]

Specifications

Color White
Assay Percent Range 99.995%
Packaging Glass bottle
Quantity 5 g
Formula Weight 334.11
Percent Purity 99.995%
Grade Trace Metal Basis
Physical Form Crystalline Powder
Chemical Name or Material Tetraammineplatinum(II) chloride hydrate
compliance-icons
Product Identifier
  • Tetraammineplatinum(II) chloride hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.