Learn More
Julabo™ Test Tube Racks for Water Bath
Specifications
Specifications
| Description | Test tube rack |
| For Use With (Equipment) | SW22, SW23, TW12, TW20, TW8 water bath |
| Holds | 50 Test Tubes, 16/17 mm dia. |
| Material | Stainless Steel |