missing translation for 'onlineSavingsMsg'
Learn More

Medline Scientific™ Test Tube Rack

Product Code. 10757384
Change view
Click to view available options
Holds:
Tubes
For Use With (Equipment):
10mm Tube Size
16mm Tube Size
25mm Tube Size
8mm Tube Size
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds For Use With (Equipment) unitSize
10757384 Tubes 25mm Tube Size Each
10599722 Tubes 16mm Tube Size Each
10041662 Tubes 10mm Tube Size Each
10286092 Tubes 8mm Tube Size Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10757384 Supplier Medline Scientific™ Supplier No. AAA23586
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with bench top and floor standing shaking incubators

The Lab Companion range of refrigerated shaking incubators include the SI-600R, a smaller version, the SI-300R and the floor model IS-971R which is a capable of incubating large volumes and very low temperatures (+4°C).
The SI-300R and SI-600R units are bench top models with a slim width to ensure they fit into the tightest of spaces. The lid, which is supported by integral gas pressure struts, opens to give front and top access for any size flask up to 2,000mL. The patented CLS control system exceeds Class II safety standards for use in areas where there may be flammable environments.
PID controller ensures accurate temperature control
  • LED display
  • Select orbital or reciprocal motion
  • Cooling coils supplied with non-refrigerated models
  • Adjustable stroke length
  • Supplied with universal platform
Range of bench top and floorstanding shaking incubators suitable for a variety of applications.

Specifications

Holds Tubes
Tube Size 25 mm dia.
Product Type Test Tube Rack
For Use With (Equipment) 25mm Tube Size
No. of Positions 19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.