Learn More
IKA™ Test Tube Attachment for IKA Platform Shaker
Product Code. 10130921 Shop All IKA Products
Change view
Click to view available options
Holds:
Tubes
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
10130921 Tubes Each
10490243 - Each
10728971 Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10130921 Supplier IKA™ Supplier No. L001850
Only null left
Add to basket
Add to basket
Attachment with customized array for holding reagent glasses on a platform shaker.
  • Requires VG 3.3 universal attachment
  • Facilitates multi-tube vortexing
TRUSTED_SUSTAINABILITY
Specifications
Holds Tubes
Tube Size 12 mm
Product Type Test Tube Adapter Attachment
Description Test tube adapter attachment
Model Model MS 1.32
For Use With (Equipment) Universal attachment, shaker
No. of Positions 6