missing translation for 'onlineSavingsMsg'
Learn More

(-)-Terpinen-4-ol, 97%, sum of enantiomers

Product Code. 10124730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10124730 100 g Glass bottle 100g
10297350 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124730 Supplier Thermo Scientific Acros Supplier No. 270431000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20126-76-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20126-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167108
InChI Key WRYLYDPHFGVWKC-JTQLQIEISA-N
Synonym --terpinen-4-ol, --4-terpineol, unii-8vi196vs5t, p-menth-1-en-4-ol, r--, r-p-menth-1-en-4-ol, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, r, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, 1r, r-1-isopropyl-4-methyl-3-cyclohexen-1-ol, l-4-terpineneol, l-4-terpineol
PubChem CID 5325830
IUPAC Name (1R)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
SMILES CC1=CCC(CC1)(C(C)C)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9330g/mL
Boiling Point 212.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (sum of enantiomers) (GC)
Packaging Glass bottle
Refractive Index 1.4760 to 1.4820
Quantity 100 g
Beilstein 06, IV, 250
Specific Gravity 0.933
Specific Rotation Condition −25° (20°C c=neat)
Specific Rotation − 25.00
Solubility Information Solubility in water: very slightly soluble., soluble in alcohol
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (-)-Terpinen-4-ol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • (-)-Terpinen-4-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.