missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR V alpha 12.1 Monoclonal Antibody (6D6.6), FITC

Product Code. 10117483
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10117483 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10117483 Supplier Invitrogen™ Supplier No. TCR2764

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TCR V alpha 12.1 Monoclonal Antibody for IHC (F), Flow, IP, TCA

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR V alpha 12.1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation
Classification Monoclonal
Clone 6D6.6
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 0.5% BSA and 0.1% sodium azide
Gene Alias TCRVA; TCRVA 12.1; V alpha 12.1; V alpha-12.1 TCR
Host Species Mouse
Immunogen Human TCR V alpha 12.1.
Purification Method Protein G
Quantity 200 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.