missing translation for 'onlineSavingsMsg'
Learn More

TC-O 9311, Tocris Bioscience™

Product Code. 18074352
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074352

Brand: Tocris Bioscience 4255/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective GPR139 agonist

Potent GPR139 agonist (EC50 = 39 nM in CHO-K1 cells expressing human GPR139). Displays no activity against a range of 90 diverse targets.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444932-31-4
Molecular Formula C20H19N3O4
Molecular Weight (g/mol) 365.39
MDL Number MFCD02372953
InChI Key KPTMSQHTGZMEFU-UHFFFAOYSA-N
Synonym 2-3,5-dimethoxybenzoyl-n-1-naphthyl hydrazinecarboxamide, 1-3,5-dimethoxybenzoyl amino-3-naphthalen-1-ylurea, n'-1-naphthyl amino carbonyl-3,5-dimethoxybenzhydrazide, 2-3,5-dimethoxyphenyl carbonyl-n-naphthalen-1-yl hydrazinecarboxamide, 3,5-dimethoxybenzoic acid 2-1-naphthalenylamino carbonyl hydrazide
PubChem CID 3376937
IUPAC Name 3,5-dimethoxy-N-{[(naphthalen-1-yl)carbamoyl]amino}benzamide
SMILES COC1=CC(=CC(OC)=C1)C(=O)NNC(=O)NC1=C2C=CC=CC2=CC=C1

Specifications

Quantity 10 mg
Formula Weight 365.38
Percent Purity >98%
Chemical Name or Material TC-O 9311
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.