missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBP Monoclonal Antibody

Description
TBP Monoclonal Antibody for ChIP, ChIP-seq, Western Blot
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
Specifications
Specifications
| Antigen | TBP |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot |
| Classification | Monoclonal |
| Concentration | 8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% thimerosal |
| Gene | Tbp |
| Gene Accession No. | P20226, P29037 |
| Gene Alias | Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100) |
| Gene Symbols | Tbp |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?