missing translation for 'onlineSavingsMsg'
Learn More

Taselisib, MedChemExpress

Product Code. 16485898
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485898 10 mg 10mg
16472047 50 mg 50mg
16468537 5 mg 5mg
16416356 100 mg 100mg
16401116 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16485898 Supplier Medchem Express Supplier No. HY13898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Taselisib (GDC-0032) is a potent PI3K inhibitor targets PIK3CA mutations, with Kis of 0.12 nM, 0.29 nM, 0.97 nM, and 9.1 nM for PI3Kδ, PI3Kα, PI3Kγ and PI3Kβ, respectively.

Taselisib (GDC-0032) is a potent PI3K inhibitor targets PIK3CA mutations, with Kis of 0.12 nM, 0.29 nM, 0.97 nM, and 9.1 nM for PI3Kδ, PI3Kα, PI3Kγ and PI3Kβ, respectively.

Specifications

Chemical Name or Material Taselisib
CAS 1282512-48-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H28N8O2
Quantity 10 mg
Synonym GDC-0032 RG-7604
Solubility Information DMSO : 50 mg/mL (108.57 mM; Need ultrasonic)
SMILES CC(C)N1C(C2=CN3C(C(C(OCC3)=C4)=CC=C4C5=CN(C(C)(C(N)=O)C)N=C5)=N2)=NC(C)=N1
Molecular Weight (g/mol) 460.53
Formula Weight 460.53
Percent Purity 99.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.