missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Protein Expression Assay Kit (hCSTB)

Product Code. 10045894 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10045894 100 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10045894 Supplier Applied Biosystems™ Supplier No. 4405465

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TaqMan™ Protein Expression Assay Kit (hCSTB)

Applied Biosystems™ TaqMan™ Protein Expression Assays enable fast, easy identification, and relative protein quantification of pluripotency protein markers from limited quantities of cultured hESCs. The TaqMan™ Protein Expression Assay Kit (Human CSTB) contains the assay probes and a dilution buffer for the labelling of human cystatin-B protein (hCSTB) in cell lysates prepared using the Protein Expression Sample Preparation Kit. The assay probes consist of two oligonucleotides conjugated to antibodies that bind to separate epitopes of the hCSTB protein. Key features of the TaqMan™ Protein Expression Assay:
  • Quantitates protein expression using gold standard TaqMan™ 5’ nuclease chemistry
  • Enables direct correlation of mRNA and/or miRNA expression to protein expression on the same real-time PCR platform
  • Allows greater sensitivity and a more quantitative readout than western blots and immunostaining
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Contains Assay Probe A, Assay Probe B, and Assay Probe Dilution Buffer. The assay probes (Assay Probe A and Assay Probe B) are supplied at a 20x concentration. The Assay Probe Dilution Buffer is supplied at a 1x concentration. Store at -15 to -25°C.
    Detection Method Primer-probe
    GC-Rich PCR Performance Low
    PCR Method qPCR
    Polymerase AmpliTaq Fast DNA Polymerase
    Reaction Speed Fast
    Technique Proximity Ligation Assay
    For Use With (Equipment) 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
    Label or Dye FAM
    Passive Reference Dye ROX (Pre-mixed)
    Product Line TaqMan
    Product Type Protein Expression Assay Kit
    Purity or Quality Grade UP (Ultra Pure)
    Quantity 100 reactions
    Shipping Condition Wet Ice
    Sufficient For 100 Reactions
    Target Specificity Target-Specific
    Hot Start No
    No. of Reactions 100 reactions
    Sample Type Cells
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.