missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Content And Storage | 1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers). Store at -15 to -25°C. |
| For Use With (Application) | Real Time PCR (qPCR) |
| Green Features | Sustainable packaging |
| No. of Reactions | 2900 reactions |
| Product Line | TaqMan |
| Quantity | L (2900 rxns), made to order |
| Shipping Condition | Room Temperature |
| Form | Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?